Please login to your online account to display your discounted pricing

anti-IL-22, Clone: 8F10C10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227321SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227321SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 50616
Concentration 0.5mg/mL
Primary or Secondary Primary
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. Q9GZX6
Monoclonal or Polyclonal Monoclonal
Quantity 0.025mg
Conjugate Unlabeled
Antigen IL-22
Isotype κ
Clone 8F10C10
Format Purified
Host Species Mouse
Immunogen MAPISSHCRL DkSNFQQPYI TNRTFMLAkE ASLADNNTDV RLIGEkLFHG VSMSERCYLM kQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQkLkDTV kkLGESGEIk AIGELDLLFM SLRNACI
Regulatory Status RUO
Cross Reactivity Human
Applications Western Blotting