Please login to your online account to display your discounted pricing

anti-IL-21, Clone: 14k5H3, Novus Biologicals™

$109.00 - $255.80

Specifications

Clone 14k5H3
Host Species Murine
Isotype κ
Immunogen Full length recombinant protein
Antigen IL-21
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227336

 
novus biologicals canada
NBP227336
Unlabeled 0.1mg Each for $255.80

NBP227336SS

 
novus biologicals canada
NBP227336SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 14k5H3
Host Species Murine
Isotype κ
Immunogen Full length recombinant protein
Antigen IL-21
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q9HBE4
Gene ID (Entrez) 59067
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL