Please login to your online account to display your discounted pricing

anti-IL-21, Clone: 14k5A9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227337

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227337

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Concentration 0.5mg/mL
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. Q9HBE4
Gene ID (Entrez) 59067
Conjugate Unlabeled
Immunogen Full length recombinant protein
Quantity 0.1mg
Antigen IL-21
Format Purified
Host Species Mouse
Regulatory Status RUO
Clone 14k5A9
Cross Reactivity Human
Applications Western Blotting