Please login to your online account to display your discounted pricing

anti-IL-21, Clone: 14k5A9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227337SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227337SS

  • / Each
 

Description & Specifications

Specifications

Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Applications Western Blotting
Gene ID (Entrez) 59067
Conjugate Unlabeled
Host Species Mouse
Immunogen Full length recombinant protein
Cross Reactivity Human
Regulatory Status RUO
Quantity 0.025mg
Format Purified
Antigen IL-21
Clone 14k5A9
Gene Accession No. Q9HBE4
Concentration 0.5mg/mL