Please login to your online account to display your discounted pricing

anti-IL-17E/IL-25, Clone: 8B1, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 8B1
Host Species Mouse
Isotype κ
Immunogen Full-length human IL17E recombinant protein was used as the immunogen for this antibody.
Antigen IL-17E/IL-25
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227324

 
novus biologicals canada
NBP227324
Unlabeled 0.1mg Each for $309.05

NBP227324SS

 
novus biologicals canada
NBP227324SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 8B1
Host Species Mouse
Isotype κ
Immunogen Full-length human IL17E recombinant protein was used as the immunogen for this antibody.
Antigen IL-17E/IL-25
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blotting
Gene Accession No. Q9H293
Gene ID (Entrez) 64806
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL