Please login to your online account to display your discounted pricing

anti-IL-17B, Clone: 97C691, Novus Biologicals™

$85.00 - $278.00

Specifications

Clone 97C691
Host Species Mouse
Isotype IgG1
Immunogen This antibody was incorrectly reported as being raised against recombinant human IL-17 BR protein. Specifically
Antigen IL-17B
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224708

 
novus biologicals canada
NBP224708
Unlabeled 0.1mg Each for $278.00

NBP224708SS

 
novus biologicals canada
NBP224708SS
Unlabeled 0.025mg Each for $85.00
Description & Specifications

Specifications

Clone 97C691
Host Species Mouse
Isotype IgG1
Immunogen This antibody was incorrectly reported as being raised against recombinant human IL-17 BR protein. Specifically
Antigen IL-17B
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity This antibody is reactive to Human.
Gene Accession No. Q9UHF5
Gene ID (Entrez) 27190
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL