Please login to your online account to display your discounted pricing

anti-IL-17B, Clone: 97C691, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP224708SS

 View all options for this product


For Research Use Only

Catalog No. NBP224708SS


Description & Specifications


Antigen IL-17B
Applications Western Blotting
Clone 97C691
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. Q9UHF5
Host Species Mouse
Immunogen This antibody was incorrectly reported as being raised against recombinant human IL-17 BR protein. Specifically
Isotype IgG1
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 27190
Test Specificity This antibody is reactive to Human.