Please login to your online account to display your discounted pricing

anti-IL-17 RA/IL-17 R, Clone: 49M5D9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225261

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225261

  • / Each
 

Description & Specifications

Specifications

Antigen IL-17 RA/IL-17 R
Applications Western Blot
Clone 49M5D9
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Gene Accession No. NP_055154
Host Species Murine
Immunogen Within the range of amino acids 186-264 of human IL-17RA were used as the immunogen for the antibody.
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 23765
Test Specificity This product reacts with Human.
Target Species Human