Please login to your online account to display your discounted pricing

anti-IL-17 RA/IL-17 R, Clone: 49M5D9, Novus Biologicals™

$139.00 - $307.00

Specifications

Clone 49M5D9
Host Species Mouse
Isotype κ
Immunogen Within the range of amino acids 186-264 of human IL-17RA were used as the immunogen for the antibody.
Antigen IL-17 RA/IL-17 R
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225261

 
novus biologicals canada
NBP225261
Unlabeled 0.1mg Each for $307.00

NBP225261SS

 
novus biologicals canada
NBP225261SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 49M5D9
Host Species Mouse
Isotype κ
Immunogen Within the range of amino acids 186-264 of human IL-17RA were used as the immunogen for the antibody.
Antigen IL-17 RA/IL-17 R
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity This product reacts with Human.
Gene Accession No. NP_055154
Gene ID (Entrez) 23765
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Concentration 0.5mg/mL