Please login to your online account to display your discounted pricing

anti-IL-17 RA/IL-17 R, Clone: 49M4D2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225258

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225258

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Isotype κ
Gene ID (Entrez) 23765
Test Specificity This product reacts with Human.
Cross Reactivity Human
Immunogen Within the range of amino acids 186-291 of human IL-17RA were used as the immunogen for the antibody.
Regulatory Status RUO
Format Purified
Host Species Mouse
Quantity 0.1mg
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen IL-17 RA/IL-17 R
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Gene Accession No. NP_055154
Primary or Secondary Primary
Clone 49M4D2
Conjugate Unlabeled