Please login to your online account to display your discounted pricing

anti-IL-17/IL-17A, Clone: 4k5F6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224765

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224765

  • / Each
 

Description & Specifications

Specifications

Antigen IL-17/IL-17A
Applications Western Blot
Clone 4k5F6
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg IL-17A antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. Q6NZ94
Host Species Murine
Immunogen A synthetic peptide between amino acids 1-75 of human IL-17A was used as the immunogen for the antibody.
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3605
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human, and 94% in Mouse, Rat, Monkey, Opossum, Horse, Cow and Chimpanzee S1PR1.
Target Species Human