Please login to your online account to display your discounted pricing

anti-IL-17/IL-17A, Clone: 4k5F6, Novus Biologicals™

$109.00 - $362.00

Specifications

Clone 4k5F6
Host Species Mouse
Isotype κ
Immunogen A synthetic peptide between amino acids 1-75 of human IL-17A was used as the immunogen for the antibody.
Antigen IL-17/IL-17A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224765

 
novus biologicals canada
NBP224765
Unlabeled 0.1mg Each for $362.00

NBP224765SS

 
novus biologicals canada
NBP224765SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 4k5F6
Host Species Mouse
Isotype κ
Immunogen A synthetic peptide between amino acids 1-75 of human IL-17A was used as the immunogen for the antibody.
Antigen IL-17/IL-17A
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human, and 94% in Mouse, Rat, Monkey, Opossum, Horse, Cow and Chimpanzee S1PR1.
Gene Accession No. Q6NZ94
Gene ID (Entrez) 3605
Formulation 100μg IL-17A antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL