Please login to your online account to display your discounted pricing

anti-IL-17/IL-17A, Biotin, Clone: 409C8.02, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0331B

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0331B

  • / Each
 

Description & Specifications

Specifications

Antigen IL-17/IL-17A
Applications Flow Cytometry
Applications Western Blot
Clone 409C8.02
Conjugate Biotin
Cross Reactivity Human
Format Ion Exchange Chromatography
Formulation Antibody in PBS 50% glycerol
Host Species Murine
Immunogen rhCytotoxic T Lymphocyte associated-Antigen 8 transfected COS-7 cells
Isotype IgG1
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3605
Target Species Human