Please login to your online account to display your discounted pricing

anti-IL-17/IL-17A, Biotin, Clone: 409C8.02, Novus Biologicals™

$406.00 - $536.00

Specifications

Clone 409C8.02
Host Species Mouse
Isotype IgG1
Immunogen rhCytotoxic T Lymphocyte associated-Antigen 8 transfected COS-7 cells
Antigen IL-17/IL-17A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX0331B

 
novus biologicals canada
DDX0331B
Biotin 0.1mg Each for $536.00

DDX0331B005

 
novus biologicals canada
DDX0331B005MG
Biotin 0.05mg Each for $406.00
Description & Specifications

Specifications

Clone 409C8.02
Host Species Mouse
Isotype IgG1
Immunogen rhCytotoxic T Lymphocyte associated-Antigen 8 transfected COS-7 cells
Antigen IL-17/IL-17A
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blotting
Gene ID (Entrez) 3605
Formulation Antibody in PBS 50% glycerol