Please login to your online account to display your discounted pricing

anti-IL-17/IL-17A, Biotin, Clone: 409C8.02, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0331B005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0331B005

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3605
Cross Reactivity Human
Format Ion Exchange Chromatography
Immunogen rhCytotoxic T Lymphocyte associated-Antigen 8 transfected COS-7 cells
Applications Flow Cytometry
Applications Western Blotting
Formulation Antibody in PBS 50% glycerol
Quantity 0.05mg
Isotype IgG1
Primary or Secondary Primary
Clone 409C8.02
Regulatory Status RUO
Antigen IL-17/IL-17A
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Conjugate Biotin