Please login to your online account to display your discounted pricing

anti-IL-1 beta/IL-1F2, Clone: 43N8F6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227341

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227341

  • / Each
 

Description & Specifications

Specifications

Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene ID (Entrez) 3553
Gene Accession No. P01584
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Immunogen Full-length recombinant human IL-1b protein was used as immunogen for this antibody.
Primary or Secondary Primary
Quantity 0.1mg
Format Purified
Host Species Mouse
Cross Reactivity Human
Applications Western Blotting
Clone 43N8F6
Regulatory Status RUO
Antigen IL-1 beta/IL-1F2