Please login to your online account to display your discounted pricing

anti-IL-1 beta/IL-1F2, Clone: 43N8F6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227341SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227341SS

  • / Each
 

Description & Specifications

Specifications

Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P01584
Immunogen Full-length recombinant human IL-1b protein was used as immunogen for this antibody.
Primary or Secondary Primary
Concentration 0.5mg/mL
Gene ID (Entrez) 3553
Cross Reactivity Human
Quantity 0.025mg
Format Purified
Applications Western Blotting
Regulatory Status RUO
Host Species Mouse
Conjugate Unlabeled
Antigen IL-1 beta/IL-1F2
Clone 43N8F6