Please login to your online account to display your discounted pricing

anti-IL-1 beta/IL-1F2, Clone: 43N4C4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227343

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227343

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3553
Concentration 0.5mg/mL
Primary or Secondary Primary
Immunogen Full-length recombinant human IL-1beta protein was used as immunogen for this antibody.
Quantity 0.1mg
Cross Reactivity Human
Regulatory Status RUO
Format Purified
Applications Western Blotting
Antigen IL-1 beta/IL-1F2
Clone 43N4C4
Gene Accession No. P01584