Please login to your online account to display your discounted pricing

anti-IL-1 beta/IL-1F2, Clone: 43N4C4, Novus Biologicals™

$109.00 - $255.80

Specifications

Clone 43N4C4
Host Species Mouse
Immunogen Full-length recombinant human IL-1beta protein was used as immunogen for this antibody.
Antigen IL-1 beta/IL-1F2
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227343SS

 
novus biologicals canada
NBP227343SS
Unlabeled 0.025mg Each for $109.00

NBP227343

 
novus biologicals canada
NBP227343
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Clone 43N4C4
Host Species Mouse
Immunogen Full-length recombinant human IL-1beta protein was used as immunogen for this antibody.
Antigen IL-1 beta/IL-1F2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. P01584
Gene ID (Entrez) 3553
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL