Please login to your online account to display your discounted pricing

anti-IL-1 beta/IL-1F2, Clone: 43N1F9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227340

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227340

  • / Each
 

Description & Specifications

Specifications

Antigen IL-1 beta/IL-1F2
Applications Western Blot
Clone 43N1F9
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. P01584
Host Species Murine
Immunogen Full-length recombinant human IL-1 beta protein was used as immunogen for this antibody.
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3553
Target Species Human