Please login to your online account to display your discounted pricing

anti-IL-1 beta/IL-1F2, Clone: 43N1F9, Novus Biologicals™

$109.00 - $255.80

Specifications

Clone 43N1F9
Host Species Murine
Isotype κ
Immunogen Full-length recombinant human IL-1 beta protein was used as immunogen for this antibody.
Antigen IL-1 beta/IL-1F2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227340

 
novus biologicals canada
NBP227340
Unlabeled 0.1mg Each for $255.80

NBP227340SS

 
novus biologicals canada
NBP227340SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 43N1F9
Host Species Murine
Isotype κ
Immunogen Full-length recombinant human IL-1 beta protein was used as immunogen for this antibody.
Antigen IL-1 beta/IL-1F2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. P01584
Gene ID (Entrez) 3553
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL