Please login to your online account to display your discounted pricing

anti-IKK gamma, Clone: 72C627, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 72C627
Host Species Murine
Isotype IgG1
Immunogen This antibody was raised against a His-tagged full-length human IKK3/IKKg protein.
Antigen IKK gamma
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056542

 
novus biologicals canada
NB10056542
Unlabeled 0.1mg Each for $329.00

NB10056542T

 
novus biologicals canada
NB10056542SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 72C627
Host Species Murine
Isotype IgG1
Immunogen This antibody was raised against a His-tagged full-length human IKK3/IKKg protein.
Antigen IKK gamma
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blot
Test Specificity A 48 kDa band is observed.
Gene Accession No. Q9Y6K9
Gene ID (Entrez) 8517
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL