Please login to your online account to display your discounted pricing

anti-IKK epsilon/IKBKE, Clone: 72B587, Novus Biologicals™

$139.00 - $321.00

Specifications

Clone 72B587
Host Species Mouse
Isotype IgG2a, κ
Immunogen This antibody was raised against a synthetic peptide corresponding to amino acids 175-188 of human IKKi/e protein.
Antigen IKK epsilon/IKBKE
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056524T

 
novus biologicals canada
NB10056524SS
Unlabeled 0.025mg Each for $139.00

NB10056524

 
novus biologicals canada
NB10056524
Unlabeled 0.1mg Each for $321.00
Description & Specifications

Specifications

Clone 72B587
Host Species Mouse
Isotype IgG2a, κ
Immunogen This antibody was raised against a synthetic peptide corresponding to amino acids 175-188 of human IKKi/e protein.
Antigen IKK epsilon/IKBKE
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Simple Western
Applications Western Blotting
Test Specificity The IKK epsilon amino acid sequence used as immunogen is 100% homologous with TBK1 (TANK-binding kinase 1).
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL