Please login to your online account to display your discounted pricing

anti-IKK epsilon/IKBKE, Clone: 72B587, Novus Biologicals™

Manufacturer: novus biologicals canada  NB10056524SS

 View all options for this product


For Research Use Only

Catalog No. NB10056524T


Description & Specifications


Antigen IKK epsilon/IKBKE
Applications Flow Cytometry
Applications Simple Western
Applications Western Blotting
Clone 72B587
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS containing 0.05% BSA
Host Species Mouse
Immunogen This antibody was raised against a synthetic peptide corresponding to amino acids 175-188 of human IKKi/e protein.
Isotype IgG2a, κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity The IKK epsilon amino acid sequence used as immunogen is 100% homologous with TBK1 (TANK-binding kinase 1).