Please login to your online account to display your discounted pricing

anti-IKK beta, Clone: 10AG2, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056509SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056509T

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Gene ID (Entrez) 3551
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Mouse
Quantity 0.025mg
Primary or Secondary Primary
Clone 10AG2
Format Purified
Regulatory Status RUO
Gene Accession No. O14920
Isotype IgG1
Test Specificity An approx. 87 kDa band should be observed.
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Immunogen A full-length human IKKb recombinant protein was used to immunize mice (NP_001547).
Monoclonal or Polyclonal Monoclonal
Antigen IKK beta