Please login to your online account to display your discounted pricing

anti-IkB-alpha (p Ser32, p Ser36), Clone: 39A1413, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056724SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056724T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 0.05% BSA
Isotype IgG1, κ
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mg
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Gene ID (Entrez) 4792
Test Specificity IkBa (39A1413 ) [Ser32/Ser36].
Gene Accession No. P25963
Antigen IkB-alpha (p Ser32
Regulatory Status RUO
Clone 39A1413
Format Purified
Host Species Mouse
Immunogen This monoclonal antibody was raised against a synthetic peptide containing phosphorylated serines at amino acid residues 32 and 36 of human IkBa.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Conjugate Unlabeled