Please login to your online account to display your discounted pricing

anti-IkB-alpha, Clone: 5D1623, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 5D1623
Host Species Mouse
Isotype IgG1
Immunogen This monoclonal antibody was developed against a synthetic peptide corresponding to the caspase-3 mediated cleavage site of IkBa.
Antigen IkB-alpha
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056597

 
novus biologicals canada
NB10056597
Unlabeled 0.1mg Each for $329.00

NB10056597T

 
novus biologicals canada
NB10056597SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 5D1623
Host Species Mouse
Isotype IgG1
Immunogen This monoclonal antibody was developed against a synthetic peptide corresponding to the caspase-3 mediated cleavage site of IkBa.
Antigen IkB-alpha
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity IkBa (5D1623).
Gene Accession No. P25963
Gene ID (Entrez) 4792
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL