Please login to your online account to display your discounted pricing

anti-IkB-alpha, Clone: 5D1623, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056597SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056597T

  • / Each
 

Description & Specifications

Specifications

Isotype IgG1
Antigen IkB-alpha
Cross Reactivity Human
Clone 5D1623
Immunogen This monoclonal antibody was developed against a synthetic peptide corresponding to the caspase-3 mediated cleavage site of IkBa.
Test Specificity IkBa (5D1623).
Gene ID (Entrez) 4792
Applications Western Blotting
Host Species Mouse
Regulatory Status RUO
Formulation PBS containing 0.05% BSA
Monoclonal or Polyclonal Monoclonal
Format Purified
Concentration 0.5mg/mL
Conjugate Unlabeled
Gene Accession No. P25963
Primary or Secondary Primary
Quantity 0.025mg