missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen IgM (Heavy chain) Cross-Adsorbed Rabbit anti-Mouse, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Supplier: Invitrogen 616800
Description
Reconstitue with 2 mL distilled water.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgM (Heavy chain) Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Mouse IgM | |
1 mg | |
Secondary | |
4° C | |
IgG |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
Rabbit | |
Affinity chromatography | |
RUO | |
Mouse | |
Lyophilized |