Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Mouse IgG1, FITC/RPE/Cy5RPE Conjugated Control

Mouse IgG1, FITC/RPE/Cy5RPE Conjugated Control from Thermo Scientific™ Pierce™ is validated for Ctrl and FACS.

$424.20 - $953.40

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PISA112476

 
pierce biotechnology inc
SA112476
FITC/RPE/Cy5RPE 50 TS Each for $424.20

PISA112475

 
pierce biotechnology inc
SA112475
FITC, RPE, RPE-Cy5 100 TS Each for $953.40
Description & Specifications

Mouse IgG1, FITC/RPE/Cy5RPE Conjugated Control from Thermo Scientific™ Pierce™ is validated for Ctrl and FACS.

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody’s species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Host Species: Mouse
Isotype: IgG1
Immunogen: White blood cells.
Formulation: 100 Tests of Affinity chromatography purified isotype control in PBS buffer, with carrier protein 0.2%;
SA1-12475 has been successfully used in Ctrl and FACS procedures.