Please login to your online account to display your discounted pricing
anti-Mouse IgG Fc, Biotin Conjugated Polyclonal, Thermo Scientific™ Pierce™ DFS Item
Mouse IgG Fc Polyclonal antibody A16088 from Thermo Scientific™ Pierce™ specifically detects Mouse IgG Fc in Mouse samples, and it is validated for IF, FACS and ELISA
$197.40 - $214.20
Products
Catalog Number Mfr. No. Price Quantity    
A16088
DFS Item
life technologies
LSA16088
Each for $197.40
A16094
DFS Item
life technologies
LSA16094
Each for $214.20
Description & Specifications
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
A16088 detects Mouse IgG Fc in Mouse samples. A16088 has been successfully used in IF, FACS and ELISA procedures. The immunogen for A16088 is gamma Immunoglobin Fc region.
Applications:
ELISA, FACS, Immunofluorescence
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.