Please login to your online account to display your discounted pricing

anti-IFN-gamma, Clone: A6C5, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0001P005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0001P005

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3458
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone A6C5
Conjugate Unlabeled
Antigen IFN-gamma
Immunogen rhuIFN gamma adsorbed with lysates of enriched Raji cell membranes
Format Ion Exchange Chromatography
Formulation Tris-NaCl pH 8
Host Species Mouse
Primary or Secondary Primary
Isotype IgG1
Quantity 0.05mg
Concentration 0.5mg/mL
Cross Reactivity Human