Please login to your online account to display your discounted pricing

anti-IFN-gamma, Clone: A6C5, Novus Biologicals™

$309.05 - $367.12

Specifications

Clone A6C5
Host Species Mouse
Isotype IgG1
Immunogen rhuIFN gamma adsorbed with lysates of enriched Raji cell membranes
Antigen IFN-gamma
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX0001P005

 
novus biologicals canada
DDX0001P005MG
Unlabeled 0.05mg Each for $309.05

DDX0001P01

 
novus biologicals canada
DDX0001P01MG
Unlabeled 0.1mg Each for $367.12

DDX0001P05

 
novus biologicals canada
DDX0001P05MG
Unlabeled 0.5mg Each for $309.05
Description & Specifications

Specifications

Clone A6C5
Host Species Mouse
Isotype IgG1
Immunogen rhuIFN gamma adsorbed with lysates of enriched Raji cell membranes
Antigen IFN-gamma
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Gene ID (Entrez) 3458
Formulation Tris-NaCl pH 8
Concentration 0.5mg/mL