Please login to your online account to display your discounted pricing

anti-IFN-gamma, Clone: A6C5, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0001P05MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0001P05

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3458
Cross Reactivity Human
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Immunogen rhuIFN gamma adsorbed with lysates of enriched Raji cell membranes
Clone A6C5
Format Ion Exchange Chromatography
Regulatory Status RUO
Host Species Mouse
Primary or Secondary Primary
Concentration 0.5mg/mL
Antigen IFN-gamma
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Formulation Tris-NaCl pH 8
Quantity 0.5mg
Isotype IgG1