Please login to your online account to display your discounted pricing

anti-IFN-gamma, Clone: A6C5, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0001P01MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0001P01

  • / Each
 

Description & Specifications

Specifications

Antigen IFN-gamma
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Clone A6C5
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Ion Exchange Chromatography
Formulation Tris-NaCl pH 8
Host Species Murine
Immunogen rhuIFN gamma adsorbed with lysates of enriched Raji cell membranes
Isotype IgG1
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3458
Target Species Human