Please login to your online account to display your discounted pricing

anti-ICAM-3/CD50, Clone: 186-2G9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329411ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329411M

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Antigen ICAM-3/CD50
Isotype κ
Host Species Mouse
Immunogen Stimulated human leukocytes
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Gene Accession No. P32942
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3385
Concentration 0.2mg/mL
Quantity 1mL
Clone 186-2G9
Regulatory Status RUO