Please login to your online account to display your discounted pricing

anti-ICAM-3/CD50, Clone: 186-2G9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232941

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232941

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Clone 186-2G9
Immunogen Stimulated human leukocytes
Quantity 0.5mL
Gene ID (Entrez) 3385
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Primary or Secondary Primary
Gene Accession No. P32942
Formulation PBS with 0.05% BSA.
Cross Reactivity Human
Antigen ICAM-3/CD50
Host Species Mouse
Regulatory Status RUO