Please login to your online account to display your discounted pricing

anti-ICAM-3/CD50, Clone: 186-2G9, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 186-2G9
Host Species Mouse
Isotype κ
Immunogen Stimulated human leukocytes
Antigen ICAM-3/CD50
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329411M

 
novus biologicals canada
NBP2329411ML
Unlabeled 1mL Each for $429.00

NBP232941

 
novus biologicals canada
NBP232941
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone 186-2G9
Host Species Mouse
Isotype κ
Immunogen Stimulated human leukocytes
Antigen ICAM-3/CD50
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P32942
Gene ID (Entrez) 3385
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL