Please login to your online account to display your discounted pricing

anti-Huntingtin, Clone: 1A771, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056703

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056703

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity Cross reacts with Human and Mouse. Not yet tested in other species.
Formulation PBS containing 0.2% gelatin
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3064
Concentration 0.5mg/mL
Antigen Huntingtin
Gene Accession No. P42858
Immunogen An Htt mutant protein containing 62 glutamine amino acid repeats was used as immunogen.
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Mouse
Quantity 0.1mg
Regulatory Status RUO
Format Purified
Applications Western Blotting
Isotype IgG
Clone 1A771