Please login to your online account to display your discounted pricing

anti-Huntingtin, Clone: 1A771, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 1A771
Host Species Murine
Isotype IgG
Immunogen An Htt mutant protein containing 62 glutamine amino acid repeats was used as immunogen.
Antigen Huntingtin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056703

 
novus biologicals canada
NB10056703
Unlabeled 0.1mg Each for $329.00

NB10056703T

 
novus biologicals canada
NB10056703SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 1A771
Host Species Murine
Isotype IgG
Immunogen An Htt mutant protein containing 62 glutamine amino acid repeats was used as immunogen.
Antigen Huntingtin
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Cross reacts with Human and Mouse. Not yet tested in other species.
Gene Accession No. P42858
Gene ID (Entrez) 3064
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL