Please login to your online account to display your discounted pricing

anti-Huntingtin, Clone: 1A771, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056703SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056703T

  • / Each
 

Description & Specifications

Specifications

Antigen Huntingtin
Applications Western Blot
Clone 1A771
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. P42858
Host Species Murine
Immunogen An Htt mutant protein containing 62 glutamine amino acid repeats was used as immunogen.
Isotype IgG
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3064
Test Specificity Cross reacts with Human and Mouse. Not yet tested in other species.
Target Species Human
Target Species Murine