Please login to your online account to display your discounted pricing

anti-Human IgG, Clone: 4D2D9G8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP151523SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP151523SS

  • / Each
 

Description & Specifications

Specifications

Antigen Human IgG
Applications ELISA
Applications Western Blot
Clone 4D2D9G8
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS
Host Species Murine
Immunogen Human IgG was isolated from human sera and purified by chromatography.
Isotype IgG1
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Secondary
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody is reactive with whole Human IgG, and has no cross reactivity with Human IgM.
Target Species Human