Please login to your online account to display your discounted pricing

anti-Human IgG, Clone: 4A11, Novus Biologicals™


Manufacturer: novus biologicals canada  NB500467

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB500467

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Western Blotting
Cross Reactivity Human
Host Species Mouse
Immunogen Full length native protein (purified) (Human).
Isotype IgG2b
Formulation PBS pH 7.4
Primary or Secondary Secondary
Concentration 1.0mg/mL
Monoclonal or Polyclonal Monoclonal
Test Specificity This reacts with Fab fragment of human IgG. Specificity was confirmed by Western blotting analysis of purified Fab fragment, of human IgG, under reducing conditions. Not yet tested in other species.
Quantity 0.1mg
Conjugate Unlabeled
Regulatory Status RUO
Antigen Human IgG
Format Purified
Clone 4A11