Please login to your online account to display your discounted pricing

anti-HSP60, Clone: LK2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232867

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232867

  • / Each
 

Description & Specifications

Specifications

Test Specificity E. coli, Borellia, Streptococcus pyogenes, Yersinia entercolitica, Salmonella typhimurium, Treponema hyodysenteriae, Treponema innocense, Trichinella spiralis, Helminths, Yeast, Spinach.
Isotype κ
Quantity 0.5mL
Cross Reactivity Bacteria
Cross Reactivity Chicken
Cross Reactivity Fungi
Cross Reactivity Guinea Pig
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P10809
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3329
Conjugate Unlabeled
Immunogen Recombinant human HSP60 protein
Host Species Mouse
Clone LK2
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL
Antigen HSP60
Regulatory Status RUO