Please login to your online account to display your discounted pricing

anti-HSP60, Clone: LK2, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone LK2
Host Species Mouse
Isotype κ
Immunogen Recombinant human HSP60 protein
Antigen HSP60
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232867

 
novus biologicals canada
NBP232867
Unlabeled 0.5mL Each for $321.00

NBP2328671M

 
novus biologicals canada
NBP2328671ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone LK2
Host Species Mouse
Isotype κ
Immunogen Recombinant human HSP60 protein
Antigen HSP60
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity E. coli, Borellia, Streptococcus pyogenes, Yersinia entercolitica, Salmonella typhimurium, Treponema hyodysenteriae, Treponema innocense, Trichinella spiralis, Helminths, Yeast, Spinach.
Gene Accession No. P10809
Gene ID (Entrez) 3329
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL