Please login to your online account to display your discounted pricing

anti-HSP60, Clone: LK1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232973

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232973

  • / Each
 

Description & Specifications

Specifications

Antigen HSP60
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone LK1
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P10809
Host Species Mouse
Immunogen Recombinant human HSP60 protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3329
Test Specificity Does not react with Bacteria, Helminths or Spinach.