Please login to your online account to display your discounted pricing

anti-HSP60, Clone: LK1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone LK1
Host Species Mouse
Isotype κ
Immunogen Recombinant human HSP60 protein
Antigen HSP60
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232973

 
novus biologicals canada
NBP232973
Unlabeled 0.5mL Each for $321.00

NBP2329731M

 
novus biologicals canada
NBP2329731ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone LK1
Host Species Mouse
Isotype κ
Immunogen Recombinant human HSP60 protein
Antigen HSP60
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Does not react with Bacteria, Helminths or Spinach.
Gene Accession No. P10809
Gene ID (Entrez) 3329
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL