Please login to your online account to display your discounted pricing

anti-HSP60, Clone: LK1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329731ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329731M

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Isotype κ
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P10809
Concentration 0.2mg/mL
Test Specificity Does not react with Bacteria, Helminths or Spinach.
Conjugate Unlabeled
Primary or Secondary Primary
Quantity 1mL
Immunogen Recombinant human HSP60 protein
Antigen HSP60
Clone LK1
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 3329
Format Purified
Host Species Mouse
Regulatory Status RUO