Please login to your online account to display your discounted pricing

anti-HSP60, Clone: GROEL730, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328681ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328681M

  • / Each
 

Description & Specifications

Specifications

Antigen HSP60
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone GROEL730
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P10809
Host Species Mouse
Immunogen Recombinant human HSP60 protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3329
Test Specificity Human. Does not react with Mouse or Rat.