Please login to your online account to display your discounted pricing

anti-HSP60, Clone: GROEL730, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232868

 View all options for this product


For Research Use Only

Catalog No. NBP232868


Description & Specifications


Antigen HSP60
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone GROEL730
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P10809
Host Species Mouse
Immunogen Recombinant human HSP60 protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3329
Test Specificity Human. Does not react with Mouse or Rat.